CH6371378720 Chem Impex International, Inc.

Ala-Pro-pNA HCl

  • MDL number MFCD00155539
  • Cas Number : 65022-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at =-4°C
Physical Form Yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-pro-pna·hcl/3087?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-pro-pna·hcl/3087?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart