CH6371378722 Chem Impex International, Inc.

Benzoyl-L-tyrosine 4-nitroanilide

  • MDL number MFCD00038109
  • Cas Number : 6154-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzoyl-l-tyrosine-4-nitroanilide/3089?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzoyl-l-tyrosine-4-nitroanilide/3089?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart