CH6371378723 Chem Impex International, Inc.

Boc-Ala-Ala-OH

  • MDL number MFCD00038564
  • Cas Number : 27317-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-alanyl-L-alanine
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-ala-ala-oh/3090?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-ala-ala-oh/3090?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart