CH6371378724 Chem Impex International, Inc.

Boc-Gly-Pro-OH

  • MDL number MFCD00057234
  • Cas Number : 14296-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-glycyl proline
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-gly-pro-oh/3092?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-gly-pro-oh/3092?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart