CH6371378727 Chem Impex International, Inc.

Gly-Asn-OH

  • MDL number MFCD00066024
  • Cas Number : 1999-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Glycyl-L-asparagine
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-asn-oh/3100?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-asn-oh/3100?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart