CH6371378730 Chem Impex International, Inc.

Gly-Ile-OH

  • MDL number MFCD00066047
  • Cas Number : 19461-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-ile-oh/3103?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-ile-oh/3103?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart