CH6371378732 Chem Impex International, Inc.

Gly-Trp-OH

  • MDL number MFCD00065978
  • Cas Number : 2390-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-trp-oh/3106?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-trp-oh/3106?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart