CH6371378734 Chem Impex International, Inc.

Ile-Tyr-OH

  • MDL number MFCD00063032
  • Cas Number : 38579-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Isoleucyl-L-tyrosine
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ile-tyr-oh/3108?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ile-tyr-oh/3108?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart