CH6371378738 Chem Impex International, Inc.

Leu-Leu-OMe�

  • MDL number MFCD00155477
  • Cas Number : 6491-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-184
Purity 97
Storage temp. Store at 0-8
Physical Form White to off-white pwoder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/leu-leu-ome-middot-hcl/3112?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/leu-leu-ome-middot-hcl/3112?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart