CH6371378740 Chem Impex International, Inc.

L-Methionine b-naphthylamide

  • MDL number MFCD00190955
  • Cas Number : 7424-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-84
Synonym L-Met-betaNA
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-methionine-beta-naphthylamide/3114?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-methionine-beta-naphthylamide/3114?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart