CH6371378744 Chem Impex International, Inc.

Pro-Gly-Gly-OH

  • MDL number MFCD00037877
  • Cas Number : 7561-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Prolylglycylglycine
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/pro-gly-gly-oh/3120?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pro-gly-gly-oh/3120?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart