CH6371378748 Chem Impex International, Inc.

L-Tryptophan b-naphthylamide

  • MDL number MFCD00051194
  • Cas Number : 3326-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Trp-betaNA
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-tryptophan-ß-naphthylamide/3125?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-tryptophan-ß-naphthylamide/3125?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart