CH6371378750 Chem Impex International, Inc.

Val-Pro-OH�

  • MDL number MFCD00191045
  • Cas Number : 105931-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/val-pro-oh-middot-hcl/3129?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/val-pro-oh-middot-hcl/3129?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart