CH6371378752 Chem Impex International, Inc.

Z-L-alanyl-L-tryptophan

  • MDL number MFCD00022761
  • Cas Number : 119645-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-alanyl-l-tryptophan/3131?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-alanyl-l-tryptophan/3131?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart