CH6371378753 Chem Impex International, Inc.

Z-Gly-D-Leu-OH

  • MDL number MFCD00155529
  • Cas Number : 57818-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-144 °C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-gly-d-leu-oh/3132?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-gly-d-leu-oh/3132?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart