CH6371378754 Chem Impex International, Inc.

Z-Gly-Phe-NH2

  • MDL number MFCD00056119
  • Cas Number : 5513-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Benzyloxycarbonylglycyl-L-phenylalanine amide
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-gly-phe-nh-sub-2-sub/3133?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-gly-phe-nh-sub-2-sub/3133?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart