CH6371378756 Chem Impex International, Inc.

Z-L-phenylalanine 4-nitroanilide

  • MDL number MFCD00038114
  • Cas Number : 19647-71-3
  • Molecular Weight : 419.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-159 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-4-nitroanilide/3137?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-4-nitroanilide/3137?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart