CH6371378761 Chem Impex International, Inc.

Fmoc-3-(2'-pyridyl)-D-alanine

  • MDL number MFCD00672565
  • Cas Number : 185379-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-165� C
Synonym Fmoc-D-Ala(2'-pyridyl)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-2-pyridyl-d-alanine/3142?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-2-pyridyl-d-alanine/3142?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart