CH6371378769 Chem Impex International, Inc.

Fmoc-(3-aminomethyl) benzoic acid

  • MDL number MFCD00273463
  • Cas Number : 155369-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-aminomethyl-benzoic-acid/3165?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-aminomethyl-benzoic-acid/3165?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart