CH6371378798 Chem Impex International, Inc.

Z-D-serine methyl ester

  • MDL number MFCD00799501
  • Cas Number : 93204-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 28-34 °C
Purity 97
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-d-serine-methyl-ester/3216?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-d-serine-methyl-ester/3216?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart