CH6371378800 Chem Impex International, Inc.

3-Amino-L-tyrosine-2HCl

  • MDL number MFCD00012964
  • Cas Number : 23279-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-158
Synonym m-NH2-L-Tyr-OH 2HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-amino-l-tyrosine-font-size-2-face-symbol-middot-font-2hcl/3218?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-amino-l-tyrosine-font-size-2-face-symbol-middot-font-2hcl/3218?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart