CH6371378801 Chem Impex International, Inc.

Boc-3-iodo-L-tyrosine

  • MDL number MFCD01632008
  • Cas Number : 71400-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-90
Synonym Boc-3-I-L-Tyr-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-iodo-l-tyrosine/3219?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-iodo-l-tyrosine/3219?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart