CH6371378810 Chem Impex International, Inc.

Fmoc-3-amino-1-carboxymethyl-pyridin-2-one

  • MDL number MFCD00673785
  • Cas Number : 204322-11-2
  • Molecular Weight : 390.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-amino-1-carboxymethyl-pyridin-2-one/3228?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-amino-1-carboxymethyl-pyridin-2-one/3228?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart