CH6371378819 Chem Impex International, Inc.

Fmoc-(3S,4S)-4-amino-3-hydroxy-pentanoic acid dicyclohexylammonium salt

  • MDL number MFCD00270200
  • Cas Number : 204316-31-4
  • Molecular Weight : 536.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-i-s-i-4-i-s-i-4-amino-3-hydroxy-pentanoic-acid-dicyclohexylammonium-salt/3237?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-i-s-i-4-i-s-i-4-amino-3-hydroxy-pentanoic-acid-dicyclohexylammonium-salt/3237?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart