CH6371378827 Chem Impex International, Inc.

Fmoc-(R,S)-2-carboxymorpholine

  • MDL number MFCD01632020
  • Cas Number : 204320-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-Cop-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-2-carboxymorpholine/3245?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-i-s-i-2-carboxymorpholine/3245?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart