CH6371378830 Chem Impex International, Inc.

Nalpha-Boc-Nbeta-allyloxycarbonyl-D-2,3-diaminopropionic acid dicyclohexylammonium salt

  • MDL number MFCD00800347
  • Cas Number : 204197-26-2
  • Molecular Weight : 469.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172-178 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-beta-sup-allyloxycarbonyl-d-2-3-diaminopropionic-acid-dicyclohexylammonium-salt/3248?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-beta-sup-allyloxycarbonyl-d-2-3-diaminopropionic-acid-dicyclohexylammonium-salt/3248?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart