CH6371378839 Chem Impex International, Inc.

Fmoc-L-glutamic acid g-[b-(5-naphthyl sulfonic acid)-ethylenediamine] ester

  • MDL number MFCD01076138
  • Cas Number : 193475-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Glu(Edans)-OH
Purity 97
Storage temp. Store at 0-8
Physical Form Amorphous white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-glutamic-acid-gamma-beta-5-naphthyl-sulfonic-acid-ethylenediamine-ester/3257?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-glutamic-acid-gamma-beta-5-naphthyl-sulfonic-acid-ethylenediamine-ester/3257?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart