CH6371378849 Chem Impex International, Inc.

Fmoc-4-methyl-D-phenylalanine

  • MDL number MFCD00270196
  • Cas Number : 204260-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167-172
Synonym Fmoc-D-Phe(4-Me)-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-methyl-d-phenylalanine/3267?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-methyl-d-phenylalanine/3267?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart