CH6371378857 Chem Impex International, Inc.

Fmoc-4-piperidino-L-proline

  • MDL number MFCD00273479
  • Cas Number : 204318-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Pro(4-piperidino)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-piperidino-l-proline/3275?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-piperidino-l-proline/3275?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart