CH6371378860 Chem Impex International, Inc.

5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine

  • MDL number MFCD06796700
  • Cas Number : 76512-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DMT-dC
Purity 99.5
Storage temp. Store at = - 4
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-2-deoxycytidine/3278?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-2-deoxycytidine/3278?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart