CH6371378866 Chem Impex International, Inc.

Fmoc-4-nitro-D-phenylalanine

  • MDL number MFCD00237029
  • Cas Number : 177966-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-217
Synonym Fmoc-D-Phe(4-NO2)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-nitro-d-phenylalanine/3284?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-nitro-d-phenylalanine/3284?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart