CH6371378884 Chem Impex International, Inc.

Z-(R,S)-3-amino-7-chloro-5-(2-chlorophenyl)-2-oxo-1,4-benzodiazepine

  • MDL number MFCD01632026
  • Cas Number : 209985-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White amorphous powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-r-i-i-s-i-3-amino-7-chloro-5-2-chlorophenyl-2-oxo-1-4-benzodiazepine/3303?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-r-i-i-s-i-3-amino-7-chloro-5-2-chlorophenyl-2-oxo-1-4-benzodiazepine/3303?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart