CH6371378885 Chem Impex International, Inc.

Z-(R,S)-3-amino-5-cyclohexyl-2-oxo-1,4-benzodiazepine

  • MDL number MFCD01074701
  • Cas Number : 146373-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White amorphous powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-r-i-i-s-i-3-amino-5-cyclohexyl-2-oxo-1-4-benzodiazepine/3304?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-r-i-i-s-i-3-amino-5-cyclohexyl-2-oxo-1-4-benzodiazepine/3304?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart