CH6371378887 Chem Impex International, Inc.

Z-(R,S)-3-amino-2-oxo-5-phenyl-1,4-benzodiazepine

  • MDL number MFCD01074700
  • Cas Number : 108895-98-3
  • Molecular Weight : 385.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-r-i-i-s-i-3-amino-2-oxo-5-phenyl-1-4-benzodiazepine/3306?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-r-i-i-s-i-3-amino-2-oxo-5-phenyl-1-4-benzodiazepine/3306?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart