CH6371378888 Chem Impex International, Inc.

Fmoc-6-piperazin-1-yl-4(3H)-quinazolinone-3-acetic acid

  • MDL number MFCD01074698
  • Cas Number : 269078-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-167 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to greyish powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-6-piperazin-1-yl-4-3-i-h-i-quinazolinone-3-acetic-acid/3307?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-6-piperazin-1-yl-4-3-i-h-i-quinazolinone-3-acetic-acid/3307?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart