CH6371378894 Chem Impex International, Inc.

Boc-piperazine hydrochloride

  • MDL number MFCD00673776
  • Cas Number : 76535-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233-237� C
Synonym Boc-Piz�
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-piperazine-hydrochloride/3313?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-piperazine-hydrochloride/3313?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart