CH6371378895 Chem Impex International, Inc.

Fmoc-piperazine hydrochloride

  • MDL number MFCD00673790
  • Cas Number : 215190-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157-161� C
Synonym Fmoc-Piz HCl
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-piperazine-hydrochloride/3314?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-piperazine-hydrochloride/3314?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart