CH6371378903 Chem Impex International, Inc.

Fmoc-L-glutamic acid g-tert-butyl ester a-amide

  • MDL number MFCD01632029
  • Cas Number : 104090-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-156
Synonym Fmoc-L-Glu(OtBu)-NH2
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-glutamic-acid-gamma-i-tert-i-butyl-ester-alpha-amide/3331?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-glutamic-acid-gamma-i-tert-i-butyl-ester-alpha-amide/3331?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart