CH6371378906 Chem Impex International, Inc.

Boc-L-leucine N,O-dimethylhydroxamide

  • MDL number MFCD00151875
  • Cas Number : 87694-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Leu-N(OMe)Me
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-leucine-i-n-i-i-o-i-dimethylhydroxamide/3335?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-leucine-i-n-i-i-o-i-dimethylhydroxamide/3335?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart