CH6371378908 Chem Impex International, Inc.

Nalpha-Fmoc-Nepsilon-trimethylsilylethoxycarbonyl-L-lysine

  • MDL number MFCD01075098
  • Cas Number : 122903-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-70 °C
Purity 99.5
Storage temp. Store at 0-8°C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-trimethylsilylethoxycarbonyl-l-lysine/3339?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-trimethylsilylethoxycarbonyl-l-lysine/3339?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart