CH6371378922 Chem Impex International, Inc.

Fmoc-alpha,beta-dehydro-2-aminobutyric acid

  • MDL number MFCD00235893
  • Cas Number : 198545-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-alpha-beta-dehydro-2-aminobutyric-acid/3355?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-alpha-beta-dehydro-2-aminobutyric-acid/3355?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart