CH6371378923 Chem Impex International, Inc.

Boc-L-a-aminobutyric acid dicyclohexylammonium salt

  • MDL number MFCD06795823

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Abu-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-alpha-aminobutyric-acid-dicyclohexylammonium-salt/3356?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-alpha-aminobutyric-acid-dicyclohexylammonium-salt/3356?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart