CH6371378926 Chem Impex International, Inc.

H-D-Bpa-OH

  • MDL number MFCD00238101
  • Cas Number : 201466-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/h-d-bpa-oh/3359?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/h-d-bpa-oh/3359?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart