CH6371378931 Chem Impex International, Inc.

Boc-L-2-cyclohexylglycine dicyclohexylammonium salt

  • MDL number MFCD01075090
  • Molecular Weight : 438.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-142 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-2-cyclohexylglycine-dicyclohexylammonium-salt/3364?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-2-cyclohexylglycine-dicyclohexylammonium-salt/3364?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart