CH6371378935 Chem Impex International, Inc.

Na,b-Bis-Fmoc-D-2,3-diaminopropionic acid

  • MDL number MFCD01632030
  • Cas Number : 1217631-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Dap(Fmoc)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-beta-sup-bis-fmoc-d-2-3-diaminopropionic-acid/3368?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-beta-sup-bis-fmoc-d-2-3-diaminopropionic-acid/3368?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart