CH6371378939 Chem Impex International, Inc.

Boc-O-benzyl-D-homoserine

  • MDL number MFCD00270496
  • Cas Number : 150009-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-57 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-homoserine/3372?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-homoserine/3372?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart