CH6371378941 Chem Impex International, Inc.

D-Homotyrosine hydrobromide salt

  • MDL number MFCD01075128
  • Cas Number : 185617-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-HomoTyr-OH HBr
Purity 95
Storage temp. Store at 0-8
Physical Form White solids
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-homotyrosine-hydrobromide-salt/3374?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-homotyrosine-hydrobromide-salt/3374?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart