CH6371378942 Chem Impex International, Inc.

Boc-L-homotyrosine dicyclohexylammonium salt

  • MDL number MFCD00235905
  • Cas Number : 126825-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-HomoTyr-OH DCHA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-homotyrosine-dicyclohexylammonium-salt/3375?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-homotyrosine-dicyclohexylammonium-salt/3375?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart