CH6371378943 Chem Impex International, Inc.

Fmoc-L-homotyrosine

  • MDL number MFCD00235904
  • Cas Number : 198560-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-homotyrosine/3376?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-homotyrosine/3376?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart