CH6371378944 Chem Impex International, Inc.

Boc-4-(2-chloro-Z-amino)-L-phenylalanine

  • MDL number MFCD01075088
  • Cas Number : 198475-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Phe(4-NH-2-CL-Z)-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-2-chloro-z-amino-l-phenylalanine/3377?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-2-chloro-z-amino-l-phenylalanine/3377?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart